Learn More
Histo Chem Incorporated 30MG FLUORO JADE C
Catalog No. NC0309912
Encompass
A Stain For Neuronal Degeneration With Fluorescent Nissl Counter Stain. For The High Contrast And Resolution Staining Of Degenerating Neurons In Formalin Fixed Brain Tissue Sections.
Catalog No. NC0309912 Supplier Histo Chem Incorporated Supplier No. 1FJC
A Stain For Neuronal Degeneration With Fluorescent Nissl Counter Stain. For The High Contrast And Resolution Staining Of Degenerating Neurons In Formalin Fixed Brain Tissue Sections.