Learn More
Chemglass Life Sciences 30mL Filter Funnel, Jacketed, Coarse Frit
SDP
Catalog No. CG140302
Encompass_Preferred
30mL Filter Funnel, Jacketed, Coarse Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.
Catalog No. CG140302 Supplier Chemglass Life Sciences Supplier No. CG140302
Add to Cart
Add to Cart
30mL Filter Funnel, Jacketed, Coarse Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.