Learn More
Bellco Glass, Inc. 30mL Centrifuge Tube
SDP
Catalog No. 502330182
Encompass_Preferred
Description30mL size is an ideal option for intermediate centrifuge tube volumesSelf-standing base for use without a rackMolded graduations ensure volume accuracy to +/- 2%Conical bottom for maximized sample retrieval Pack Description50/bagCase Quantity500SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume30mLGraduatedYesGraduation Increments (Subdivisions)5mLMax Force (RCF)g7500RNase/DNase FreeYes
Catalog No. 50-233-0182 Supplier Bellco Glass, Inc. Supplier No. 229494000
Description30mL size is an ideal option for intermediate centrifuge tube volumesSelf-standing base for use without a rackMolded graduations ensure volume accuracy to +/- 2%Conical bottom for maximized sample retrieval Pack Description50/bagCase Quantity500SterileYesNon-pyrogenicYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume30mLGraduatedYesGraduation Increments (Subdivisions)5mLMax Force (RCF)g7500RNase/DNase FreeYes