Learn More
Bellco Glass, Inc. 30mL Graduated Beaker, Polypropylene, Non-sterile
SDP
Catalog No. 502330371
Encompass_Preferred
The Polypropylene graduated beaker has molded  pouring spouts and rolled up lip for drip-less work environment. They have angle walls for easier handling and sample retrieving. Nesting design for easy stacking and storage.\n\nSpecifications:\n
    \n
  • Smaller case quantities are easier to store and are ideal for small scale users.
  • \n
  • Polypropylene material fir resistance to a variety of strong acids and solvents.
  • \n
  • Size:30mL
  • \n
  • Pack Description: 100/Bag
  • \n
  • Case Quantity: 100
  • \n
Catalog No. 50-233-0371 Supplier Bellco Glass, Inc. Supplier No. 230511000
The Polypropylene graduated beaker has molded  pouring spouts and rolled up lip for drip-less work environment. They have angle walls for easier handling and sample retrieving. Nesting design for easy stacking and storage.\n\nSpecifications:\n
    \n
  • Smaller case quantities are easier to store and are ideal for small scale users.
  • \n
  • Polypropylene material fir resistance to a variety of strong acids and solvents.
  • \n
  • Size:30mL
  • \n
  • Pack Description: 100/Bag
  • \n
  • Case Quantity: 100
  • \n