Learn More
CELLTREAT Scientific Products 30mL Oak Ridge Style Round Bottom Centrifuge Tube, Non-sterile
SDP
Catalog No. 50828731
Encompass_Preferred
CELLTREAT Scientific Products 30mL Oak Ridge Centrifuge Tube. High speed tubes feature a round bottom and uniform wall thickness. Tubes are economical and leak-proof for refrigerated or standard centrifugation. For proper performance, 80% of total capacity must be filled. Max force (RCF) g 50000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 12/case.
Catalog No. 50-828-731 Supplier CELLTREAT Scientific Products Supplier No. 229461
CELLTREAT Scientific Products 30mL Oak Ridge Centrifuge Tube. High speed tubes feature a round bottom and uniform wall thickness. Tubes are economical and leak-proof for refrigerated or standard centrifugation. For proper performance, 80% of total capacity must be filled. Max force (RCF) g 50000. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 12/case.