Learn More
Ace Glass, Inc. 30ML PESTLE ONLY 30ML PESTLE ONLY
SDP
Catalog No. 50979773
Encompass_Preferred
30mL glass Pestle with 10mm rod, for use with 30mL tapered tube (50974570) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.
Catalog No. 50-979-773 Supplier Ace Glass, Inc. Supplier No. 832524
30mL glass Pestle with 10mm rod, for use with 30mL tapered tube (50974570) as a tissue grinder. The rod can be used with a 509558293 Flex-Grip Chuck.