Learn More
Ace Glass, Inc. 30mL Gooch, high form crucible, with 30mm diameter porosity A (145-174 micron) fritted disc. Height above the fritted disc is 45mm.
SDP
Catalog No. 50974145
Encompass_Preferred
30mL Gooch, high form crucible, with 30mm diameter porosity A (145-174 micron) fritted disc. Height above the fritted disc is 45mm.
Catalog No. 50-974-145 Supplier Ace Glass, Inc. Supplier No. 717002
30mL Gooch, high form crucible, with 30mm diameter porosity A (145-174 micron) fritted disc. Height above the fritted disc is 45mm.