Learn More
Ace Glass, Inc. 30mL Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
SDP
Supplier:  Ace Glass, Inc. 718622
Encompass_Preferred
30mL Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
Catalog No. 50-979-165