Learn More
Ace Glass, Inc. 30mL Gooch, high form crucible, with 30mm diameter porosity C (25-50 micron) fritted disc. Height above the fritted disc is 45mm.
SDP
Catalog No. 50974147
Encompass_Preferred
30mL Gooch, high form crucible, with 30mm diameter porosity C (25-50 micron) fritted disc. Height above the fritted disc is 45mm.
Catalog No. 50-974-147 Supplier Ace Glass, Inc. Supplier No. 717006
30mL Gooch, high form crucible, with 30mm diameter porosity C (25-50 micron) fritted disc. Height above the fritted disc is 45mm.