Learn More
Ace Glass, Inc. 30mL Gooch, high form crucible, with 30mm diameter porosity D (10-20 micron) fritted disc. Height above the fritted disc is 45mm.
SDP
Catalog No. 50979122
Encompass_Preferred
30mL Gooch, high form crucible, with 30mm diameter porosity D (10-20 micron) fritted disc. Height above the fritted disc is 45mm.
Catalog No. 50-979-122 Supplier Ace Glass, Inc. Supplier No. 717008
30mL Gooch, high form crucible, with 30mm diameter porosity D (10-20 micron) fritted disc. Height above the fritted disc is 45mm.