Learn More
Alkali Scientific 30mL Centrifgue Tubes, PP, Free Standing, Flat screw cap, 25/Sterile bag
SDP
Catalog No. 502389383
Encompass_Preferred
30mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free
Catalog No. 50-238-9383 Supplier Alkali Scientific Supplier No. C5630
30mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free