Learn More
CEM Corporation 30 mL Reaction Vessel Assembly
SDP
Catalog No. 501877668
Encompass
The 30 mL reaction vessel assembly includes the reaction vessel, union, and associated fittings and ferrules. It should be used for synthetic scales less than 0.5 mmol.
Catalog No. 50-187-7668 Supplier CEM Corporation Supplier No. 542435
The 30 mL reaction vessel assembly includes the reaction vessel, union, and associated fittings and ferrules. It should be used for synthetic scales less than 0.5 mmol.