Learn More
Chemglass Life Sciences Funnel, 30mL, Separatory, Blank, Stopper Neck, Size # 13, Glassblowers
SDP
Catalog No. CG62401
Encompass_Preferred
Funnel, 30mL, Separatory, Blank, Stopper Neck, Size # 13, Glassblowers. Squibb shaped separatory blank complete with tooled and finished ground stopper neck top. Lower portion of blank is unfinished and requires a stopcock to be sealed. NOTE: SUPPLIED WITHOUT STOPPER.
Catalog No. CG62401 Supplier Chemglass Life Sciences Supplier No. CG62401
Funnel, 30mL, Separatory, Blank, Stopper Neck, Size # 13, Glassblowers. Squibb shaped separatory blank complete with tooled and finished ground stopper neck top. Lower portion of blank is unfinished and requires a stopcock to be sealed. NOTE: SUPPLIED WITHOUT STOPPER.