Learn More
Beckman Coulter Sterile and Certified Free Tubes
Supplier:  Beckman Coulter C14307
Encompass
The Beckman Coulter Sterile and Certified Free Tubes are sterile, pyrogen-free, and certified DNAse, RNAse, and PCR inhibitor free. They are made of polypropylene and come in a variety of sizes from 0.5mL to 50mL. The tubes have uniformly shaped conical bottoms and leakproof, screw caps. They are suitable for sample collection, storage, processing, and analysis in clinical, pharmaceutical, food, environmental, and research labs. The tubes provide reliable containment and are compatible with most lab equipment and analyzers.
Catalog No. NC1877945