Learn More
Savillex Corporation 30 mL Standard Vial, Rounded Interior
SDP
Catalog No. NC1563235
Encompass
Flat surface on bottom of vial ideal for hot plate heating . Round interior allows for easy removal of contents. Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C).
Catalog No. NC1563235 Supplier Savillex Corporation Supplier No. 20003020
Flat surface on bottom of vial ideal for hot plate heating . Round interior allows for easy removal of contents. Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C).