Learn More
Ace Glass, Inc. 30ML VOLUMETRIC BULB 30ML VOLUMETRIC BULB
SDP
Catalog No. 50979613
Encompass_Preferred
30mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-979-613 Supplier Ace Glass, Inc. Supplier No. 800439
30mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.