Learn More
Chemglass Life Sciences 10" Diameter Upper Agitator, PTFE, Improved
SDP
Catalog No. 501531446
Encompass_Preferred
10" Diameter Upper Agitator, PTFE, Improved. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 30mm OD stirrer shaft CG-2097 in 75 and 100L reactors. The 45 angle impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. The upper agitator hub is open so that it can be positioned wherever the indents are along the 30mm shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. Heavy-duty blades are 6.5mm thick. PLEASE NOTE: THE LOWER AGITATOR HAS BUSHING TO ACCEPT A STABILIZING PIN FROM CG-1968-Q-02. THE PIN EXTENDS INTO THE LOWER AGITATOR AND VIRTUALLY ELIMINATES ANY WHIPPING BY STABILIZING THE STIRRER SHAFT.
Catalog No. 50-153-1446 Supplier Chemglass Life Sciences Supplier No. CG2095R50
10" Diameter Upper Agitator, PTFE, Improved. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 30mm OD stirrer shaft CG-2097 in 75 and 100L reactors. The 45 angle impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. The upper agitator hub is open so that it can be positioned wherever the indents are along the 30mm shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. Heavy-duty blades are 6.5mm thick. PLEASE NOTE: THE LOWER AGITATOR HAS BUSHING TO ACCEPT A STABILIZING PIN FROM CG-1968-Q-02. THE PIN EXTENDS INTO THE LOWER AGITATOR AND VIRTUALLY ELIMINATES ANY WHIPPING BY STABILIZING THE STIRRER SHAFT.