Learn More
THORLABS INC 30 mm Cage Cube-Mounted Non-Polarizing Beamsplitter, 1100 - 1600 nm, 8-32 Tap
Catalog No. NC2152238
Encompass
30 mm Cage Cube-Mounted Non-Polarizing Beamsplitter, 1100 - 1600 nm, 8-32 Tap
This item is not returnable. View return policy
Catalog No. NC2152238 Supplier THORLABS INC Supplier No. CCM1BS015
30 mm Cage Cube-Mounted Non-Polarizing Beamsplitter, 1100 - 1600 nm, 8-32 Tap