Learn More
Azzota Corp 30mm fluor cuvette 10.5ml 4win
Catalog No. 5000886897
Encompass_Preferred
45x12 5x32 5mm 30mm Pathlength Standard Fluorimeter Cuvettes 10 5ml Glass 340nm-2500nm Four polished windows
Catalog No. 50-008-86897 Supplier Azzota Corp Supplier No. G206
45x12 5x32 5mm 30mm Pathlength Standard Fluorimeter Cuvettes 10 5ml Glass 340nm-2500nm Four polished windows