Learn More
Ace Glass, Inc. 30MM GRADED SEALS 30MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845540
Encompass
30mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-784