Learn More
Azzota Corp 30mm Path Fluorometer 10.5ml Q
Catalog No. 5000887035
Encompass_Preferred
45x12 5x32 5mm 30mm Pathlength Standard Fluorimeter Cuvettes 10 5ml Quartz 190nm-2500nm Four polished windows
Catalog No. 50-008-87035 Supplier Azzota Corp Supplier No. Q206
45x12 5x32 5mm 30mm Pathlength Standard Fluorimeter Cuvettes 10 5ml Quartz 190nm-2500nm Four polished windows