Learn More
Ace Glass, Inc. 30MM POR C FILTER FUNNEL 30Mm Por C Filter Funnel
SDP
Supplier:  Ace Glass, Inc. 718506
Encompass
Funnel with jacket for cooling or heating. All porosities of a given size are priced the same. Hose connection is size C.
Catalog No. 50-979-161