Learn More
Ace Glass, Inc. 30MM POR C FUNNEL 30MM POR C FUNNEL
SDP
Catalog No. 50979174
Encompass_Preferred
30mm porosity C (25-50 micron) filter funnel with a 60 degree angle. Funnel is useful where it is necessary to wash the precipitation and re-dissolve with chemicals that would attach filter paper.
Catalog No. 50-979-174 Supplier Ace Glass, Inc. Supplier No. 718716
Add to Cart
Add to Cart
30mm porosity C (25-50 micron) filter funnel with a 60 degree angle. Funnel is useful where it is necessary to wash the precipitation and re-dissolve with chemicals that would attach filter paper.