Learn More
Ace Glass, Inc. 30MM POR D FUNNEL 30MM POR D FUNNEL
SDP
Catalog No. 50974304
Encompass_Preferred
30mm porosity D (10-20 micron) filter funnel with a 60 degree angle. Funnel is useful where it is necessary to wash the precipitation and re-dissolve with chemicals that would attach filter paper.
Catalog No. 50-974-304 Supplier Ace Glass, Inc. Supplier No. 718718
30mm porosity D (10-20 micron) filter funnel with a 60 degree angle. Funnel is useful where it is necessary to wash the precipitation and re-dissolve with chemicals that would attach filter paper.