Learn More
Chemglass Life Sciences 30mm Quartz Fritted Disc, Medium
SDP
Catalog No. CGQ020713
Encompass_Preferred
30mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020713 Supplier Chemglass Life Sciences Supplier No. CGQ020713
30mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.