Learn More
Chemglass Life Sciences Optional 30mm Stirrer Shaft, Reactor Size: 100L, Fits Support Frame: CG-1968-F-24 & CG-1968-F-26, Overall Length: 58.8"
SDP
Catalog No. CG2097100
Encompass_Preferred
30mm Stirrer Shaft, Reactor Size: 100L, Fits Support Frame: CG-1968-F-24 & CG-1968-F-26, Overall Length: 58.8". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. PLEASE NOTE: CG- CG-2097-100 IS MADE FROM HEAVY WALL TUBING AND HAS INDENTS FOR AGITATOR POSITIONING. INDENTS ARE SPACED EVERY 10¿ FROM THE BOTTOM AGITATOR.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. CG2097100 Supplier Chemglass Life Sciences Supplier No. CG2097100
30mm Stirrer Shaft, Reactor Size: 100L, Fits Support Frame: CG-1968-F-24 & CG-1968-F-26, Overall Length: 58.8". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. PLEASE NOTE: CG- CG-2097-100 IS MADE FROM HEAVY WALL TUBING AND HAS INDENTS FOR AGITATOR POSITIONING. INDENTS ARE SPACED EVERY 10¿ FROM THE BOTTOM AGITATOR.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.