Learn More
Chemglass Life Sciences Chemglass 30 mm Hollow Stirrer Shaft for 100 L Process Reactor (CG-2097-102)
SDP
Catalog No. NC1982865
Encompass_Preferred
The Chemglass CG-2097-102 is a precision 30 mm hollow stirrer shaft designed for use with 100 L process reactors. The shaft is precision ground and polished, with the lower portion sandblasted and a 3/8 in flat for secure mounting. It is made from heavy-wall glass tubing and is compatible with PTFE agitators and reactor support frames.
  • 30 mm outer diameter
  • Hollow shaft construction
  • Lower portion sandblasted with 3/8 in flat
  • Designed for 100 L process reactors
  • Precision ground and polished finish
  • Made from heavy-wall glass tubing
  • Compatible with PTFE agitators and reactor support frames
Catalog No. NC1982865 Supplier Chemglass Life Sciences Supplier No. CG2097102
The Chemglass CG-2097-102 is a precision 30 mm hollow stirrer shaft designed for use with 100 L process reactors. The shaft is precision ground and polished, with the lower portion sandblasted and a 3/8 in flat for secure mounting. It is made from heavy-wall glass tubing and is compatible with PTFE agitators and reactor support frames.
  • 30 mm outer diameter
  • Hollow shaft construction
  • Lower portion sandblasted with 3/8 in flat
  • Designed for 100 L process reactors
  • Precision ground and polished finish
  • Made from heavy-wall glass tubing
  • Compatible with PTFE agitators and reactor support frames