Learn More
Chemglass Life Sciences 30mm Stirrer Shaft Coupling, For Use With CG-2033-X-11 Motor, Gold Motor Adapter
SDP
Catalog No. 5000980597
Encompass_Preferred
Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench. PLEASE NOTE: OTHER THANCG-2097 PROCESS REACTOR STIRRER SHAFTS, CHEMGLASS 19MM OD STIRRER SHAFTS HAVE A REDUCED TOP DIAMETER OF 12. 7MM. WHEN ORDERING A UNIVERSAL COUPLING FOR A CHEMGLASS 19MM OD STIRRER SHAFT, USE PART NUMBER CG-2046-03.
Catalog No. 50-009-80597 Supplier Chemglass Life Sciences Supplier No. CG204697
Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench. PLEASE NOTE: OTHER THANCG-2097 PROCESS REACTOR STIRRER SHAFTS, CHEMGLASS 19MM OD STIRRER SHAFTS HAVE A REDUCED TOP DIAMETER OF 12. 7MM. WHEN ORDERING A UNIVERSAL COUPLING FOR A CHEMGLASS 19MM OD STIRRER SHAFT, USE PART NUMBER CG-2046-03.