Learn More
Chemglass Life Sciences 30mm Stirrer Shaft, Reactor Size: 150L, Fits Support Frame: CG-1968-Z-150 & CG-1968-Z-151, Overall Length: 63"
SDP
Catalog No. 5000980619
Encompass_Preferred
30mm polished, hollow stirrer shaft with a solid top for mounting. Lower portion is sandblasted and has two indents for mounting 10” O. D. PTFE Agitators.
Catalog No. 50-009-80619 Supplier Chemglass Life Sciences Supplier No. CG2097X150
30mm polished, hollow stirrer shaft with a solid top for mounting. Lower portion is sandblasted and has two indents for mounting 10” O. D. PTFE Agitators.