Learn More
Chemglass Life Sciences 30mm Stirrer Shaft Coupling, Universal, 3/8" OD Stud
SDP
Catalog No. 501531417
Encompass_Preferred
30mm Stirrer Shaft Coupling, Universal, 3/8" OD Stud. Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench.
Catalog No. 50-153-1417 Supplier Chemglass Life Sciences Supplier No. CG204606
30mm Stirrer Shaft Coupling, Universal, 3/8" OD Stud. Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench.