Learn More
Bellco Glass, Inc. 30mm x 10mm Glass Bottom Tissue Culture Treated Dish, 15mm Glass, Sterile
SDP
Catalog No. 502329587
Encompass_Preferred
Recessed, consistent borosilicate glass bottom is optically clear, and provides micro well ideal for cell culture and microscopy or imagingNon-cytotoxic medical adhesive does not inhibit cell growth30mm Dish has offset bottom for improved handling30mm Dish lid has molded spacers to allow gas exchange and a stacking ring for increased stabilityGlass bottom is #1.5 thickness Pack Description10/Re-sealable BagCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialDish: PolystyreneSecondary MaterialBottom: GlassWorking Volume1.4-3.5mLTissue Culture TreatedYesCell Growth Area1.77cm2RNase/DNase FreeYes
Catalog No. 50-232-9587 Supplier Bellco Glass, Inc. Supplier No. 229632000
Recessed, consistent borosilicate glass bottom is optically clear, and provides micro well ideal for cell culture and microscopy or imagingNon-cytotoxic medical adhesive does not inhibit cell growth30mm Dish has offset bottom for improved handling30mm Dish lid has molded spacers to allow gas exchange and a stacking ring for increased stabilityGlass bottom is #1.5 thickness Pack Description10/Re-sealable BagCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialDish: PolystyreneSecondary MaterialBottom: GlassWorking Volume1.4-3.5mLTissue Culture TreatedYesCell Growth Area1.77cm2RNase/DNase FreeYes