Learn More
Biotang Inc 30-Place Cardboard Slide Tray, 40/cs, Features thumb cut for easy slide removal, the horizontal compartments are recessed to protect the specimens when the cover is closed.
Catalog No. 507514986 Shop All BIOTANG Inc Products
Encompass_Preferred
30-Place Cardboard Slide Tray, 40/cs, Features thumb cut for easy slide removal, the horizontal compartments are recessed to protect the specimens when the cover is closed.
Catalog No. 50-751-4986 Supplier Biotang Inc Supplier No. HS9930
30-Place Cardboard Slide Tray, 40/cs, Features thumb cut for easy slide removal, the horizontal compartments are recessed to protect the specimens when the cover is closed.