Learn More
Medicus Health 30 Place, Stainless Steel Slide Staining Rack with Handle
SDP
Catalog No. NC2114566
Encompass
∙ 30 Place Stainless Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned ∙ Holds slides size 3"L x 1"W, 1.5"W, 2"W ∙ 30 Place Stainless Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides ∙ Great Hematology Departments who need to do Wright's Stains ∙ Slide Staining Dipper is sold by the each ∙ Slide Dipper measures: 5″W x 4″D x 3.4″H ∙ Material: Stainless steel
This item is not returnable. View return policy
Catalog No. NC2114566 Supplier Medicus Health Supplier No. 5441M3
∙ 30 Place Stainless Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned ∙ Holds slides size 3"L x 1"W, 1.5"W, 2"W ∙ 30 Place Stainless Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides ∙ Great Hematology Departments who need to do Wright's Stains ∙ Slide Staining Dipper is sold by the each ∙ Slide Dipper measures: 5″W x 4″D x 3.4″H ∙ Material: Stainless steel