Learn More
Medicus Health Rolling Specimen Transport Cooler with Biohazard Symbol
SDP
Catalog No. NC3364386
Encompass
Insulated Rolling Specimen Transport Tote Cooler with Embroidered Biohazard Symbol has a 30 Quart capacity, telescoping handle, durable roller wheels and an embroidered orange biohazard symbol on the bag for proper specimen transport marking
∙ 14"W x 11"D x 14"H (37"H extended handle height)
∙ Equipped with a top zippered main compartment with a leak-resistant and heat-sealed PEVA internal liner that's water resistant and easy to clean internal
∙ Can be used to transport frozen, refrigerated or room temperature specimens
∙ Rremovable cooler floor so it conveniently folds up making it easy to store (collapsed dimensions: 14"H x 7"D x 14"W)
∙ Includes front zippered pocket that's perfect for required lab specimen documentation
Specimen Courier Tote features a wheeled base and a telescopic handle for easy transport
∙ Also designed with two side handles to make it easy to load into the car or carry
Catalog No. NC3364386 Supplier Medicus Health Supplier No. 7049M5
Insulated Rolling Specimen Transport Tote Cooler with Embroidered Biohazard Symbol has a 30 Quart capacity, telescoping handle, durable roller wheels and an embroidered orange biohazard symbol on the bag for proper specimen transport marking
∙ 14"W x 11"D x 14"H (37"H extended handle height)
∙ Equipped with a top zippered main compartment with a leak-resistant and heat-sealed PEVA internal liner that's water resistant and easy to clean internal
∙ Can be used to transport frozen, refrigerated or room temperature specimens
∙ Rremovable cooler floor so it conveniently folds up making it easy to store (collapsed dimensions: 14"H x 7"D x 14"W)
∙ Includes front zippered pocket that's perfect for required lab specimen documentation
Specimen Courier Tote features a wheeled base and a telescopic handle for easy transport
∙ Also designed with two side handles to make it easy to load into the car or carry