Learn More
Chemglass Life Sciences Chemglass 30 μL Automation Clear Tips for Agilent Systems, Nest (Sterile), 384 tips/rack, 10 racks/pack
SDP
Catalog No. 502733225
Encompass_Preferred
Chemglass 30 μL automation clear tips for Agilent systems are sterile, nest-packed pipette tips made from medical-grade polypropylene for automated liquid handling. They are designed for high-throughput workflows and feature low-retention surfaces, DNase/RNase-free certification, and electron-beam sterilization.
  • 30 μL tip volume.
  • Clear, medical-grade polypropylene construction.
  • Sterile and DNase/RNase free.
  • Electron-beam sterilized for aseptic workflows.
  • Low-retention surface to reduce sample carryover.
  • Supplied 384 tips per rack; 10 racks per pack; 50 racks per case.
  • Produced in a controlled clean-room environment.
Catalog No. 50-273-3225 Supplier Chemglass Life Sciences Supplier No. CLS3499A701
Add to Cart
Add to Cart
Chemglass 30 μL automation clear tips for Agilent systems are sterile, nest-packed pipette tips made from medical-grade polypropylene for automated liquid handling. They are designed for high-throughput workflows and feature low-retention surfaces, DNase/RNase-free certification, and electron-beam sterilization.
  • 30 μL tip volume.
  • Clear, medical-grade polypropylene construction.
  • Sterile and DNase/RNase free.
  • Electron-beam sterilized for aseptic workflows.
  • Low-retention surface to reduce sample carryover.
  • Supplied 384 tips per rack; 10 racks per pack; 50 racks per case.
  • Produced in a controlled clean-room environment.