Learn More
Bellco Glass, Inc. 30µL Pipette, MLA, Precision, Silver
SDP
Catalog No. 502329850
Encompass_Preferred
30µL Pipette, MLA, Precision, SilverTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorSilverPrimary MaterialAluminum Alloy & Stainless SteelVolume30 μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±3.00%Precision0.93%
Catalog No. 50-232-9850 Supplier Bellco Glass, Inc. Supplier No. 451900013
30µL Pipette, MLA, Precision, SilverTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorSilverPrimary MaterialAluminum Alloy & Stainless SteelVolume30 μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±3.00%Precision0.93%