Learn More
CELLTREAT Scientific Products 30mm x 10mm Glass Bottom Tissue Culture Treated Dish, 15mm Glass, Sterile
SDP
Catalog No. 501149057
Encompass_Preferred
CELLTREAT Scientific Products 30mm x 10mm Tissue Culture Treated Dish Glass Bottom. Recessed, consistent borosilicate glass bottom is optically clear, and provides micro well ideal for cell culture and microscopy or imaging. Non-cytotoxic medical adhesive does not inhibit cell growth. 30mm Dish has offset bottom for improved handling. 30mm Dish lid has molded spacers to allow gas exchange and a stacking ring for increased stability. 15mm glass bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 50/case.
Catalog No. 50-114-9057 Supplier CELLTREAT Scientific Products Supplier No. 229632
CELLTREAT Scientific Products 30mm x 10mm Tissue Culture Treated Dish Glass Bottom. Recessed, consistent borosilicate glass bottom is optically clear, and provides micro well ideal for cell culture and microscopy or imaging. Non-cytotoxic medical adhesive does not inhibit cell growth. 30mm Dish has offset bottom for improved handling. 30mm Dish lid has molded spacers to allow gas exchange and a stacking ring for increased stability. 15mm glass bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 50/case.