Learn More
PRODUCTION BASICS ASSEMBLY STATION, 30"D x 60"W
Catalog No. NC1207647
Encompass_Preferred
The Assembly Station starts with our RTW Table and is finished with a hardwood butcher block worksurface. A hardwood worksurface makes this assembly bench extremely robust, performing well under heavy loads and repeated impact. Use this workbench for heavy-duty applications such as packaging, shipping and assembly departments. It is also suitable where handling of metal products or products with sharp edges or where frequent repositioning is required. To add accessories, attach 48” or 60” Uprights.
Catalog No. NC1207647 Supplier PRODUCTION BASICS Supplier No. 1766
The Assembly Station starts with our RTW Table and is finished with a hardwood butcher block worksurface. A hardwood worksurface makes this assembly bench extremely robust, performing well under heavy loads and repeated impact. Use this workbench for heavy-duty applications such as packaging, shipping and assembly departments. It is also suitable where handling of metal products or products with sharp edges or where frequent repositioning is required. To add accessories, attach 48” or 60” Uprights.