Learn More
Justrite™ 31 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
Shop All Justrite Safety Group Products
Description
- Under Fume Hood Cabinets are fully compliant and designed for the safe storage and venting of solvents and flammable liquids
- Single-door cabinets are offered in either left or right-hinge door closures for easy operation where space is at a premium
- Back wall vents with built-in flame arresters are positioned at lower left and top right for jurisdictions that mandate venting of vapors
- Designed to meet OSHA and NFPA standards
- Constructed of sturdy 18-gauge (1-mm) thick double-wall, welded steel with 1-1/2 in. (38-mm) of insulating air space for fire resistance
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof 2 in. (51-mm) bottom sump. They adjust on 3 in. (76-mm) centers for versatile storage. Cabinets also feature dual vents with flame arresters, four adjustable self-leveling feet, grounding connector, and trilingual warning label. Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
- Manual-close door(s) opens to a full 180 degrees and self-latches when pushed closed
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Door Style | Manual-close |
| Load Bearing Capacity | 31 gal. |
| Color | Silver |