Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 31-Norlanostenol | 16910-39-7 | 1 MG
SDP

Catalog No. 5000418841
Encompass_Preferred

31-Norlanostenol is a triterpenic compound isolated from the latex of Euphorbia officinarum. It can act as an efficient insect growth regulator on S. frugiperda and Tenebrio molitor. This product is for research use only.

  • Triterpenic compound
  • Isolated from the latex of Euphorbia officinarum
  • Acts as an efficient insect growth regulator on S. frugiperda and Tenebrio molitor
  • Purity: 99.94%
  • Molecular Weight: 414.71
  • Formula: C29H50O
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants, Euphorbiaceae, Euphorbia boetica

Catalog No. 50-004-18841 Supplier Medchemexpress LLC Supplier No. HYN72671MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

31-Norlanostenol is a triterpenic compound isolated from the latex of Euphorbia officinarum. It can act as an efficient insect growth regulator on S. frugiperda and Tenebrio molitor. This product is for research use only.

  • Triterpenic compound
  • Isolated from the latex of Euphorbia officinarum
  • Acts as an efficient insect growth regulator on S. frugiperda and Tenebrio molitor
  • Purity: 99.94%
  • Molecular Weight: 414.71
  • Formula: C29H50O
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants, Euphorbiaceae, Euphorbia boetica

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.