Learn More
Ace Glass, Inc. Rotameter tube, size 31, with stainless steel float:5.0 to45mL/min air flow, 0.065-0.65 mL/min water flow, glass float not supplied
SDP
Supplier:  Ace Glass, Inc. 748115
Encompass
Catalog No. 50-974-812