Learn More
Ace Glass, Inc. Vacuum manifold, 310mm long, (3) stopcock ports with 24/40inner joints
SDP
Catalog No. 50979843
Encompass_Preferred
310mm long, single tube vacuum manifold featuring 3 stopcock ports with 24/40 inner joints. Distance between ports is 95mm which allows for use of flasks up to 3L.
Catalog No. 50-979-843 Supplier Ace Glass, Inc. Supplier No. 874512
310mm long, single tube vacuum manifold featuring 3 stopcock ports with 24/40 inner joints. Distance between ports is 95mm which allows for use of flasks up to 3L.