Learn More
Ace Glass, Inc. Vacuum manifold, 310mm long, (3) stopcock ports with #15inner joints
SDP
Supplier:  Ace Glass, Inc. 874514
Encompass_Preferred
310mm long, single tube vacuum manifold featuring 3 stopcock ports with #15 o-ring joints. Distance between ports is 95mm which allows for use of flasks up to 3L.
Catalog No. 50-976-311