Learn More
Quality Lab Accessories Stainless Steel Spring Style Sinker, 316 Stainless Steel
Catalog No. NC9180758
Encompass
Capsule Sinker, Type 316 stainless steel, 6 spiral type, 19.3mm x 7mm, Sinkers are available coated or uncoated, PTFE coated sinkers work with magnetic retrievers whilst uncoated type 316 stainless steel sinkers are more durable, 1/EA
Catalog No. NC9180758 Supplier Quality Lab Accessories Supplier No. CAPWST19
Capsule Sinker, Type 316 stainless steel, 6 spiral type, 19.3mm x 7mm, Sinkers are available coated or uncoated, PTFE coated sinkers work with magnetic retrievers whilst uncoated type 316 stainless steel sinkers are more durable, 1/EA