Learn More
New Jersey Fluid Systems LLC 316 SS SANITARY FITTING 1/4IN
Catalog No. NC1889408
Encompass
316 SS Sanitary Fitting 1/4in Tube OD x 1/2in Kwik-Clamp Flange
Catalog No. NC1889408 Supplier New Jersey Fluid Systems LLC Supplier No. SS400SC8
316 SS Sanitary Fitting 1/4in Tube OD x 1/2in Kwik-Clamp Flange