Learn More
McMaster-Carr 316 Stainless Steel Threaded Pipe Fitting
Catalog No. NC1474856
Encompass
316 Stainless Steel Threaded Pipe FittingLow-Pressure, Bushing Adapter, 3/8 Male x 1/8 Female
Catalog No. NC1474856 Supplier McMaster-Carr Supplier No. 4452K162
316 Stainless Steel Threaded Pipe FittingLow-Pressure, Bushing Adapter, 3/8 Male x 1/8 Female