Learn More
Biosurface Technologies Corp Stainless Steel (316L) Disc Coupon, 0.5 inch diameter for use with CDC, IBR, and RDR Biofilm Reactors
Catalog No. NC9492516
Encompass
This 0.5 inch (12.7mm) diameter coupon is manufactured from 316L stainless steel, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 7 per supplier requirements.
Catalog No. NC9492516 Supplier Biosurface Technologies Corp Supplier No. RD128316
This 0.5 inch (12.7mm) diameter coupon is manufactured from 316L stainless steel, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 7 per supplier requirements.