Learn More
Eagle Stainless Container Eagle Stainless, 20 Liter Container with Lid, Gasket and Lock Lever Ring, 316L Stainless Steel. (CTL Series)
SDP
Catalog No. NC9591799
Encompass_Preferred
20 Liter container with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC9591799 Supplier Eagle Stainless Container Supplier No. CTL30316LJ
20 Liter container with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.