Learn More
Eagle Stainless Container Eagle Stainless, Stability Testing Bottle, 25ml, 316L Stainless Steel. (STB Series)
SDP
Catalog No. NC1029864
Encompass_Preferred
25ml Stability Testing Bottle with 1.5" tri-clamp opening, type 316L stainless steel and electropolished finish. Supplied with 1.5" stainless steel endcap, 1.5" platinum cured silicone gasket and 1.5" heavy duty clamp. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC1029864 Supplier Eagle Stainless Container Supplier No. STBEP25316LA
25ml Stability Testing Bottle with 1.5" tri-clamp opening, type 316L stainless steel and electropolished finish. Supplied with 1.5" stainless steel endcap, 1.5" platinum cured silicone gasket and 1.5" heavy duty clamp. Pharmaceutical grade finish of 10 Ra or better.