Learn More
Analytical Sales & Services 316L Stainless Steel Electrode. No holes.
Catalog No. 5000805141
Encompass
316L Stainless Steel Electrode (no holes)\nfor use as anode in Undivided Flow Cell\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each\n
Catalog No. 50-008-05141 Supplier Analytical Sales & Services Supplier No. F1088309
316L Stainless Steel Electrode (no holes)\nfor use as anode in Undivided Flow Cell\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each\n